ENA292595915 Enamine

2-(1,2-dimethyl-1H-imidazole-4-sulfonamido)acetic acid

  • MDL number MFCD16039994
  • Cas Number : 1235439-62-9
  • Molecular Weight : 233.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-58949

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-58949
Availability details for
Quantity
  • Availability
  • Add to Cart