ENA292595916 Enamine

N'-cyclopropyl(dodecylsulfanyl)methanimidamide hydrobromide

  • Cas Number : 1235441-79-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HBr
Product Link https://www.enaminestore.com/catalog/EN300-59564

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-59564
Availability details for
Quantity
  • Availability
  • Add to Cart