ENA292595920 Enamine

4-[methyl(propan-2-yl)amino]benzoic acid hydrochloride

  • MDL number MFCD16039999
  • Cas Number : 1235439-72-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-59612

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-59612
Availability details for
Quantity
  • Availability
  • Add to Cart