ENA292595921 Enamine

1-[2-(2,5-dimethylpyrrolidin-1-yl)ethyl]piperazine

  • MDL number MFCD16040000
  • Cas Number : 1235439-57-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-59627

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-59627
Availability details for
Quantity
  • Availability
  • Add to Cart