ENA292595923 Enamine

4-chloro-1,1-dimethyl-1H,3H-furo[3,4-c]pyridin-3-one

  • MDL number MFCD16040003
  • Cas Number : 285991-71-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 158...160
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-60612

Safety Information

msds https://www.enaminestore.com/catalog/EN300-60612
Availability details for
Quantity
  • Availability
  • Add to Cart