ENA292595924 Enamine

1-methanesulfonylcyclopentane-1-carboxylic acid

  • MDL number MFCD16040005
  • Cas Number : 1235439-64-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-60831

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-60831
Availability details for
Quantity
  • Availability
  • Add to Cart