ENA292595926 Enamine

1-({1,3-dimethyl-1H-pyrazolo[3,4-b]pyridin-5-yl}sulfanyl)-N,N-dimethylformamide

  • MDL number MFCD16040007
  • Cas Number : 1235439-63-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 144...146
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-60870

Safety Information

msds https://www.enaminestore.com/catalog/EN300-60870
Availability details for
Quantity
  • Availability
  • Add to Cart