ENA292595928 Enamine

[1,3-bis(4-bromophenyl)propan-2-yl](methyl)amine hydrochloride

  • MDL number MFCD16040009
  • Cas Number : 1235438-89-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-60970

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-60970
Availability details for
Quantity
  • Availability
  • Add to Cart