ENA292595932 Enamine

N-(pentan-3-yl)-3,4-dihydro-2H-1,4-benzoxazine-2-carboxamide hydrochloride

  • MDL number MFCD16040012
  • Cas Number : 1235438-84-2
  • Molecular Weight : 284.79

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61174

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-61174
Availability details for
Quantity
  • Availability
  • Add to Cart