ENA292595933 Enamine

(4-fluorophenyl)(2-iodophenyl)methanamine hydrochloride

  • MDL number MFCD16040013
  • Cas Number : 1235440-02-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61225

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-61225
Availability details for
Quantity
  • Availability
  • Add to Cart