ENA292595934 Enamine

3-methyl-1-phenyl-4-(trifluoromethyl)-1H-pyrazol-5-amine

  • MDL number MFCD16040014
  • Cas Number : 1235438-91-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61243

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-61243
Availability details for
Quantity
  • Availability
  • Add to Cart