ENA292595935 Enamine

2-(piperazin-1-yl)-1,3-benzothiazole dihydrochloride

  • MDL number MFCD16040016
  • Cas Number : 928026-66-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 298...300
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61333

Safety Information

msds https://www.enaminestore.com/catalog/EN300-61333
Availability details for
Quantity
  • Availability
  • Add to Cart