ENA292595940 Enamine

[(5-bromo-2-fluorophenyl)methyl]hydrazine hydrochloride

  • MDL number MFCD16040021
  • Cas Number : 1235440-14-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 140...142
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61397

Safety Information

msds https://www.enaminestore.com/catalog/EN300-61397
Availability details for
Quantity
  • Availability
  • Add to Cart