ENA292595944 Enamine

N-(cyclopropylmethyl)-1,3-thiazolidine-4-carboxamide hydrochloride

  • MDL number MFCD16040025
  • Cas Number : 1251923-95-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61457

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-61457
Availability details for
Quantity
  • Availability
  • Add to Cart