ENA292595945 Enamine

N-methyl-N-phenylpiperidine-4-carboxamide hydrochloride

  • MDL number MFCD16040026
  • Cas Number : 1235440-15-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-61460
Availability details for
Quantity
  • Availability
  • Add to Cart