ENA292595948 Enamine

N-methyl-N-(propan-2-yl)-1,3-thiazolidine-4-carboxamide hydrochloride

  • MDL number MFCD16040029
  • Cas Number : 1251923-66-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61463

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-61463
Availability details for
Quantity
  • Availability
  • Add to Cart