ENA292595952 Enamine

4-(1H-1,3-benzodiazol-1-yl)benzene-1-carboximidamide hydrochloride

  • MDL number MFCD16040033
  • Cas Number : 1235441-72-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61474

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-61474
Availability details for
Quantity
  • Availability
  • Add to Cart