ENA292595953 Enamine

1-(3,3-diethoxypropyl)-2,5-dimethyl-1H-pyrrole

  • MDL number MFCD16040035
  • Cas Number : 1235441-43-6
  • Molecular Weight : 225.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61476

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-61476
Availability details for
Quantity
  • Availability
  • Add to Cart