ENA292595954 Enamine

N-(3-aminopropyl)thiophene-2-carboxamide hydrochloride

  • MDL number MFCD03161177
  • Cas Number : 1235440-89-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 197...199
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61484

Safety Information

msds https://www.enaminestore.com/catalog/EN300-61484
Availability details for
Quantity
  • Availability
  • Add to Cart