ENA292595961 Enamine

4-(pyrrolidine-1-carbonyl)-1,3-thiazolidine hydrochloride

  • MDL number MFCD16040042
  • Cas Number : 1251923-58-6
  • Molecular Weight : 222.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61525

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-61525
Availability details for
Quantity
  • Availability
  • Add to Cart