ENA292595962 Enamine

N-cyclopropyl-N-methyl-1,3-thiazolidine-4-carboxamide hydrochloride

  • MDL number MFCD16040043
  • Cas Number : 1251923-36-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61526

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-61526
Availability details for
Quantity
  • Availability
  • Add to Cart