ENA292595963 Enamine

4-chloro-2-(4-ethylpiperazine-1-carbonyl)phenol hydrochloride

  • MDL number MFCD16040044
  • Cas Number : 1235438-79-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61528

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-61528
Availability details for
Quantity
  • Availability
  • Add to Cart