ENA292595965 Enamine

2-chloro-4-fluoro-6-(trifluoromethyl)aniline

  • MDL number MFCD16040046
  • Cas Number : 1235439-54-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61547

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-61547
Availability details for
Quantity
  • Availability
  • Add to Cart