ENA292595971 Enamine

5-ethyl-6-hydroxy-3-(pyridin-3-yl)-2-sulfanyl-3,4-dihydropyrimidin-4-one

  • MDL number MFCD16040052
  • Cas Number : 1235438-83-1
  • Molecular Weight : 249.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61587

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-61587
Availability details for
Quantity
  • Availability
  • Add to Cart