ENA292595972 Enamine

2-(5-methyl-2-phenyl-1,3-oxazol-4-yl)ethan-1-amine hydrochloride

  • MDL number MFCD16040054
  • Cas Number : 501031-56-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61591

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-61591
Availability details for
Quantity
  • Availability
  • Add to Cart