ENA292595974 Enamine

methyl 2-(dimethoxymethyl)-3-methylbutanoate

  • MDL number MFCD16040056
  • Cas Number : 143770-73-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61602

Safety Information

msds https://www.enaminestore.com/catalog/EN300-61602
Availability details for
Quantity
  • Availability
  • Add to Cart