ENA292595975 Enamine

N-[5-(aminomethyl)-2-methoxyphenyl]acetamide

  • MDL number MFCD16040057
  • Cas Number : 1235440-86-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 115...117
Purity 95.00
Storage temp. 0
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61620

Safety Information

msds https://www.enaminestore.com/catalog/EN300-61620
Availability details for
Quantity
  • Availability
  • Add to Cart