ENA292595977 Enamine

[(5-bromo-2-fluorophenyl)methyl](methyl)amine hydrochloride

  • MDL number MFCD16040059
  • Cas Number : 1235439-04-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 197...199
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61627

Safety Information

msds https://www.enaminestore.com/catalog/EN300-61627
Availability details for
Quantity
  • Availability
  • Add to Cart