ENA292595978 Enamine

[(5-bromo-2-fluorophenyl)methyl](ethyl)amine hydrochloride

  • MDL number MFCD16040060
  • Cas Number : 1235439-66-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Product Link https://www.enaminestore.com/catalog/EN300-61629

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-61629
Availability details for
Quantity
  • Availability
  • Add to Cart