ENA292595979 Enamine

(3-bromo-4,5-dimethoxyphenyl)methanamine hydrochloride

  • MDL number MFCD16040061
  • Cas Number : 1235440-00-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Product Link https://www.enaminestore.com/catalog/EN300-61632

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-61632
Availability details for
Quantity
  • Availability
  • Add to Cart