ENA292595980 Enamine

1-(cyclopropylcarbamoyl)cyclopropane-1-carboxylic acid

  • MDL number MFCD16040062
  • Cas Number : 1235441-18-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 145...147
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61636

Safety Information

msds https://www.enaminestore.com/catalog/EN300-61636
Availability details for
Quantity
  • Availability
  • Add to Cart