ENA292595985 Enamine

5-methyl-2-(piperidin-3-yl)-1H-1,3-benzodiazole dihydrochloride

  • MDL number MFCD07781106
  • Cas Number : 1235440-11-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 296...298
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61670

Safety Information

msds https://www.enaminestore.com/catalog/EN300-61670
Availability details for
Quantity
  • Availability
  • Add to Cart