ENA292595987 Enamine

3-(4-fluorophenyl)cyclobutane-1-carboxylic acid

  • MDL number MFCD16040068
  • Cas Number : 1235439-86-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 81-84
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61702

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-61702
Availability details for
Quantity
  • Availability
  • Add to Cart