ENA292595988 Enamine

1-(6-amino-1,2,3,4-tetrahydroquinolin-1-yl)ethan-1-one hydrochloride

  • MDL number MFCD16040070
  • Cas Number : 1235441-74-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 209...211
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61719

Safety Information

msds https://www.enaminestore.com/catalog/EN300-61719
Availability details for
Quantity
  • Availability
  • Add to Cart