ENA292595989 Enamine

N-methyl-N-propyl-1,3-thiazolidine-4-carboxamide hydrochloride

  • MDL number MFCD16040071
  • Cas Number : 1251924-29-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 126...128
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61721

Safety Information

msds https://www.enaminestore.com/catalog/EN300-61721
Availability details for
Quantity
  • Availability
  • Add to Cart