ENA292595992 Enamine

N-[4-(2-chloropropanamido)phenyl]thiophene-2-carboxamide

  • MDL number MFCD16040074
  • Cas Number : 1235441-48-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-61727

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-61727
Availability details for
Quantity
  • Availability
  • Add to Cart