ENA292595994 Enamine

N-(2-aminoethyl)-2H-indazole-3-carboxamide hydrochloride

  • MDL number MFCD16040076
  • Cas Number : 1235440-50-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61733

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-61733
Availability details for
Quantity
  • Availability
  • Add to Cart