ENA292595996 Enamine

4'-fluoro-1',2'-dihydrospiro[cyclopentane-1,3'-indole]

  • MDL number MFCD16040078
  • Cas Number : 1235439-61-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61747

Safety Information

msds https://www.enaminestore.com/catalog/EN300-61747
Availability details for
Quantity
  • Availability
  • Add to Cart