ENA292596000 Enamine

3-hydroxy-4-(piperazin-1-yl)-1lambda6-thiolane-1,1-dione dihydrochloride

  • MDL number MFCD16040082
  • Cas Number : 1235441-10-7
  • Molecular Weight : 293.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61775

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-61775
Availability details for
Quantity
  • Availability
  • Add to Cart