ENA292596006 Enamine

N-(2-aminoethyl)cyclopentanecarboxamide hydrochloride

  • MDL number MFCD16040088
  • Cas Number : 1235439-28-7
  • Molecular Weight : 192.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-61794

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-61794
Availability details for
Quantity
  • Availability
  • Add to Cart