ENA292596007 Enamine

4-(aminomethyl)pyridin-2-ol dihydrobromide

  • MDL number MFCD16040089
  • Cas Number : 1235439-27-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 239...241
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61806

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-61806
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart