ENA292596008 Enamine

2-cyclohexylpropan-2-amine hydrochloride

  • MDL number MFCD16040090
  • Cas Number : 90726-02-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 258...260
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61810

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-61810
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart