ENA292596013 Enamine

tert-butyl 4-(4-aminobenzoyloxy)piperidine-1-carboxylate

  • MDL number MFCD16040095
  • Cas Number : 1235441-68-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61840

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-61840
Availability details for
Quantity
  • Availability
  • Add to Cart