ENA292596018 Enamine

N-[2-(piperazin-1-yl)ethyl]morpholine-4-sulfonamide dihydrochloride

  • MDL number MFCD16040101
  • Cas Number : 1235441-61-8
  • Molecular Weight : 351.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-61871

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-61871
Availability details for
Quantity
  • Availability
  • Add to Cart