ENA292596023 Enamine

5-ethyl-6-hydroxy-3-(3-methoxyphenyl)-2-sulfanyl-3,4-dihydropyrimidin-4-one

  • MDL number MFCD16040106
  • Cas Number : 1235440-68-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61911

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-61911
Availability details for
Quantity
  • Availability
  • Add to Cart