ENA292596024 Enamine

N-(2-aminoethyl)-2,2-dimethylpropanamide hydrochloride

  • MDL number MFCD16040107
  • Cas Number : 1235441-01-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61916

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-61916
Availability details for
Quantity
  • Availability
  • Add to Cart