ENA292596025 Enamine

tert-butyl 2-[(4-aminobenzoyloxy)methyl]pyrrolidine-1-carboxylate

  • MDL number MFCD16040108
  • Cas Number : 1235439-65-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61928

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-61928
Availability details for
Quantity
  • Availability
  • Add to Cart