ENA292596027 Enamine

1-(4-aminopiperidin-1-yl)butan-1-one hydrochloride

  • MDL number MFCD16040110
  • Cas Number : 1235440-40-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61958

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-61958
Availability details for
Quantity
  • Availability
  • Add to Cart