ENA292596028 Enamine

2-amino-2-(2-chloro-6-fluorophenyl)acetonitrile hydrochloride

  • MDL number MFCD16040111
  • Cas Number : 1235439-69-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61959

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-61959
Availability details for
Quantity
  • Availability
  • Add to Cart