ENA292596030 Enamine

N,N-dimethyl-1-({2-methylpyrazolo[1,5-a]pyrimidin-6-yl}sulfanyl)formamide

  • MDL number MFCD16040113
  • Cas Number : 1235441-56-1
  • Molecular Weight : 236.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-62015

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-62015
Availability details for
Quantity
  • Availability
  • Add to Cart